missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Accessories for RT Basic Series and RT Touch Series Magnetic Stirrers

Product Code. p-6094305 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
170 mm
220 mm
Type:
Plate Cover
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15228904

Brand: Thermo Scientific™ 88880145

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these accessories to optimize performance of your Thermo Scientific™ RT Basic Series Magnetic Stirrers.

Use these Thermo Scientific™ Accessories for RT Basic Series Magnetic Stirrers to optimize performance of your stirrer.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Diameter (Metric) 170 mm
For Use With (Equipment) RT Basic Series and RT Touch Series Magnetic Stirrers
Holds 96-Well Plates, 384-Well Plates
Type Plate Cover
Material Silicone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.