Learn More
Cytiva Accessories and Replacement Parts for Amersham™ VacuGene XL Vacuum Blotting System
Product Code. 10488824 Shop All Cytiva Products
Change view
Click to view available options
Type:
Gel Supporting Screen
Masks
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10488824 Gel Supporting Screen Each
10321275 Masks Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10488824 Supplier Cytiva Supplier No. 18101043
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
VacuGene XL vacuum blotting system is the choice for nucleic acid blotting.
  • Rapidly transfer DNA from agarose gels up to 20 x 30cm in less than 1 hour
  • Efficient and reproducible transfers improve blotting results
  • Carry out all pretransfer steps in the unit
  • Precise vacuum control for optimal transfer conditions
  • RNA transfers in less than 4 hours
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Amersham VacuGene XL Vacuum Blotting System
Type Gel Supporting Screen