missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Replacement Chart Recorder Paper
Shop All Thermo Scientific Products
Click to view available options
Installation:
Customer Installed
Unit Size:
Each
Description
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
Specifications
Specifications
| Description | Chart paper for single pen chart recorders |
| For Use With (Equipment) | MaxQ™ Single Pen Chart Recorders |
| Lighting | LED |
| Temperature Range (Metric) | 0°C to 60°C |
| Type | Replacement Chart Recorder Paper |
| Installation | Customer Installed |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction