Learn More
Agilent BioTek Accessories For MultiFlo Microplate Dispenser - Manifolds
Accessories for MultiFlo Microplate Dispenser
Brand:  Agilent BioTek 1260534S
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 16512112
  • £870.00 / Pack of 48
Estimated Shipment: 10-07-2024
to see stock.
Add to basket
Description
Description
  • Syringe manifold
  • Available with 6, 24, or 48-well plates
  • Autoclavable
Specifications
Specifications
Autoclavable
48-well