Learn More
Thermo Scientific™ Accessories for Cimarec+™ Stirrer Series
Product Code. 15453660 Shop All Thermo Scientific Products
Click to view available options
Type:
Clamp and holder
PT1000 External probe 0-400C
Supporting Rod
Transparent shield for 10x10
Transparent shield for 4x4
Transparent shield for 7x7
Unit Size:
Each
Product Code. 15453660
Brand: Thermo Scientific™ CIC0001446
Only null left
Add to basket
Add to basket
Get precise stirring control for your routine protocols.
These accessories are for use with the Cimarec+™ series digital stirrers. Our new Thermo Scientific™ Cimarec+™ series digital stirrers offer precise stirring control for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operation.
Advanced technology
  • Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strongmagnetic coupling
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for stirring
  • Raised display design protects electronics from spills
  • Optional splash guard protection shield
  • Variable sizes for different sample volumes
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Display LED
Type Transparent shield for 10x10