Learn More
Thermo Scientific™ Access Port (38 mm), Customer Install
Product Code. 15533781 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric) Inside:
38 mm
missing translation for 'unitSize'
Each
Product Code. 15533781
missing translation for 'mfr': Thermo Scientific™ 505101
Only null left
Add to basket
Add to basket
For Use With: Environmental Chambers and Incubators
Provide a point of entry for probes and monitoring devices in environmental chambers and incubators with Access Ports.
TRUSTED_SUSTAINABILITY
Specifications
Description Access Port
Lighting LED
Type Access Port
Installation Customer, on center right wall
Diameter (Metric) Inside 38 mm