Learn More
BINDER™ Right Access Port (10 to 30 mm)
Product Code. 15642056 Shop All BINDER Products
Click to view available options
Material:
Steel
Unit Size:
Each
Product Code. 15642056
Brand: BINDER™ 80121017
Only null left
Add to basket
Add to basket
For Use With: BINDER units ED 56, ED 115, ED 260, ED 720
  • For use on right
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 30 mm, right
For Use With (Equipment) BINDER Chambers ED 56, ED 115, ED 260, ED 720
Material Steel
Type Access Port
Includes Silicone plug
Diameter (Metric) Inside 30 mm