Learn More
BINDER™ Access Port
For Use With: MK 240
Brand: BINDER™ 8012-1850
Additional Details : Weight : 0.00200kg
Description
- Notch-type
- For use in door
- For use with Dynamic Climate Chambers
Specifications
Notch-type access port - notch-type access port in door, 100 x 35 mm | |
BINDER Chambers MK 240 | |
Access Port | |
100 x 35 mm |