Learn More
BINDER™ Access Port
For Use With: MKFT 240
Brand:  BINDER™ 8012-1857
Additional Details : Weight : 0.00200kg
Product Code. 15913032
  • £2658.00 / Each
Estimated Shipment: 16-08-2024
to see stock.
Add to basket
Description
Description
  • Notch-type
  • For use in door
  • For use with Dynamic Climate Chambers
Specifications
Specifications
Notch-type access port - notch-type access port in door, 100 x 35 mm
BINDER Chambers MKFT 240
Access Port
100 x 35 mm