missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBP Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£188.00 - £375.00
Specifications
| Antigen | ACBP |
|---|---|
| Applications | Western Blot, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18368784
|
Novus Biologicals
NBP3-03241-20UL |
20 μL |
£188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18352696
|
Novus Biologicals
NBP3-03241-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACBP Polyclonal specifically detects ACBP in Human samples. It is validated for Western Blot, Knockout Validated.Specifications
| ACBP | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| acyl-Coenzyme A binding domain containing 1, acyl-Coenzyme A bindingprotein), CCK-RP, cholecystokinin-releasing peptide, trypsin-sensitive, diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein), Diazepam-binding inhibitor, endozepine, EP, GABA receptor modulator, MGC70414 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-114 of human ACBP (NP_001171513.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS with 50% glycerol, pH7.3. | |
| 1622 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title