missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBD7 Antibody (005), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90286AF750
This item is not returnable.
View return policy
Description
ACBD7 Monoclonal specifically detects ACBD7 in Human samples. It is validated for ELISA.
Specifications
| ACBD7 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 414149 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 5 | |
| ELISA | |
| acyl-CoA binding domain containing 7, acyl-CoA-binding domain-containing protein 7, acyl-Coenzyme A binding domain containing 7, bA455B2.2, FLJ38219, FLJ52263, MGC33893 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ACBD7 (Q8N6N7; Met1-Ile88). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction