missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACBD6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580295
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ACBD6.
ACBD6 binds long-chain acyl-coenzyme A molecules with a strong preference for unsaturated C18:1-CoA, lower affinity for unsaturated C20:4-CoA, and very weak affinity for saturated C16:0-CoA. Does not bind fatty acids.
Specifications
| ACBD6 | |
| Polyclonal | |
| Unconjugated | |
| ACBD6 | |
| 0610010G04Rik; 2610100E10Rik; Acbd6; acyl-CoA binding domain containing 6; acyl-CoA-binding domain-containing protein 6; acyl-Coenzyme A binding domain containing 6 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 84320 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9BR61 | |
| ACBD6 | |
| Recombinant Human ACBD6 protein (Met1-Ala282). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction