missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACBD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568079
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Acyl-CoA binding protein which acts as the peroxisome receptor for pexophagy but is dispensable for aggrephagy and nonselective autophagy. Binds medium- and long-chain acyl-CoA esters.
Specifications
| ACBD5 | |
| Polyclonal | |
| Unconjugated | |
| ACBD5 | |
| 1300014E15Rik; Acbd5; acyl-CoA binding domain containing 5; acyl-CoA-binding domain-containing protein 5; acyl-Coenzyme A binding domain containing 5; endozepine-related protein; KIAA1996; membrane-associated diazepam binding inhibitor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 307170, 74159, 91452 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3 | |
| A0FKI7, Q5T8D3, Q5XG73 | |
| ACBD5 | |
| Synthesized peptide derived from human ACBD5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction