missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACAT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568620
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
This gene represents a member of the 'frizzled' gene family, which encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The protein encoded by this family member contains a signal peptide, a cysteine-rich domain in the N-terminal extracellular region, and seven transmembrane domains, but unlike other family members, this protein does not contain a C-terminal PDZ domain-binding motif. This protein functions as a negative regulator of the canonical Wnt/beta-catenin signaling cascade, thereby inhibiting the processes that trigger oncogenic transformation, cell proliferation, and inhibition of apoptosis. Alternative splicing results in multiple transcript variants.
Specifications
| ACAT2 | |
| Polyclonal | |
| Unconjugated | |
| Acat2 | |
| Ab2-076; Acat2; Acat3; acetoacetyl Coenzyme A thiolase; acetyl-CoA acetyltransferase 2; acetyl-CoA acetyltransferase, cytosolic; Acetyl-CoA transferase-like protein; acetyl-Coenzyme A acetyltransferase 2; acetyl-Coenzyme A acetyltransferase 2 (acetoacetyl Coenzyme A thiolase); acetyl-Coenzyme A acetyltransferase 3; ACTL; AE1; AW742799; BND3; CD233; cytosolic acetoacetyl-CoA thiolase; DI; EMPB3; EPB3; FR; RTA1A; SW; t-complex protein 1, related sequence 1; Tcp1-rs1; Tcp-1x; WD; WD1; WR | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 39 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BWD1 | |
| Acat2 | |
| synthetic peptide directed towards the N terminal region of human ACAT2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Del en indholdskorrektion