missing translation for 'onlineSavingsMsg'
Learn More

ACAT2 Antibody, FITC, Novus Biologicals™

Product Code. 18431962 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431962 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431962 Supplier Novus Biologicals Supplier No. NBP232198F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ACAT2 Polyclonal antibody specifically detects ACAT2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAT2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias acetoacetyl Coenzyme A thiolase, acetyl-CoA acetyltransferase 2, acetyl-CoA acetyltransferase, cytosolic, Acetyl-CoA transferase-like protein, acetyl-Coenzyme A acetyltransferase 2, acetyl-Coenzyme A acetyltransferase 2 (acetoacetyl Coenzyme A thiolase), ACTL, Cytosolic acetoacetyl-CoA thiolase, EC 2.3.1, EC 2.3.1.9
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 150 to 200 of human Acetyl-CoA Acetyltransferase 2 using the numbering given in entry NP_005882.2 (GeneID 39).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 39
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.