missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACAT1 Recombinant Rabbit Monoclonal Antibody (23GB1255), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554132
This item is not returnable.
View return policy
Description
ACAT-1 is a ~50 kDa protein that catalyzes the formation of intracellular cholesterol esters in various tissues, such as adult human liver, adrenal glands, macrophages, and kidneys.
Specifications
| ACAT1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Acat1 | |
| 6330585C21Rik; Acat; Acat1; acetoacetyl Coenzyme A thiolase; acetoacetyl-CoA thiolase; acetyl-Co A acetyltransferase 1 mitochondrial; acetyl-CoA acetyltransferase 1; acetyl-CoA acetyltransferase, mitochondrial; acetyl-Coenzyme A acetyltransferase 1; MAT; mitochondrial acetoacetyl-CoA thiolase; RATACAL; T2; testicular tissue protein Li 198; THIL | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25014, 38 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1255 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17764, P24752 | |
| Acat1 | |
| A synthesized peptide derived from human ACAT1 (390-427AA). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction