missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACAT1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519227
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
ACAT-1 is a ~50 kDa protein that catalyzes the formation of intracellular cholesterol esters in various tissues, such as adult human liver, adrenal glands, macrophages, and kidneys.
Specifications
| ACAT1 | |
| Polyclonal | |
| Unconjugated | |
| Acat1 | |
| 6330585C21Rik; Acat; Acat1; acetoacetyl Coenzyme A thiolase; acetoacetyl-CoA thiolase; acetyl-Co A acetyltransferase 1 mitochondrial; acetyl-CoA acetyltransferase 1; acetyl-CoA acetyltransferase, mitochondrial; acetyl-Coenzyme A acetyltransferase 1; MAT; mitochondrial acetoacetyl-CoA thiolase; RATACAL; T2; testicular tissue protein Li 198; THIL | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 110446, 25014, 38 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P17764, P24752, Q8QZT1 | |
| Acat1 | |
| Synthetic peptide sequence (DEEYKRVDFSKVPK) corresponding to the internal amino acids of ACAT1 (aa 253-266). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction