missing translation for 'onlineSavingsMsg'
Learn More

ACAT1 Antibody (2C5), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18424965 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424965 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424965 Supplier Novus Biologicals Supplier No. NBP222565MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ACAT1 Monoclonal antibody specifically detects ACAT1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAT1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2C5
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ACAT, acetoacetyl Coenzyme A thiolase, Acetoacetyl-CoA thiolase, acetyl-CoA acetyltransferase 1, acetyl-CoA acetyltransferase, mitochondrial, acetyl-Coenzyme A acetyltransferase 1, EC 2.3.1, EC 2.3.1.9, MAT, mitochondrial acetoacetyl-CoA thiolase, T2, THIL
Host Species Mouse
Immunogen Recombinant human ACAT1 (34-427aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Lipid and Metabolism, Mitochondrial Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 38
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.