missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Acarbose

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Inhibitor of intestinal α-glucosidase (IC50 = 11 nM). Antidiabetic; inhibits the hydrolysis of complex carbohydrates.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 645.25 |
| Purity | >98% |
| Molecular Weight (g/mol) | 645.608 |
| Chemical Name or Material | Acarbose |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction