missing translation for 'onlineSavingsMsg'
Learn More

Acarbose, MedChemExpress

Product Code. 16481366
Encompass_Preferred
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
This item is not returnable. View return policy

Product Code. 16481366

Brand: Medchem Express HYB0089

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Acarbose (BAY g 5421), antihyperglycemic agent, is an orally active alpha-glucosidase inhibitor (IC50=11 nM). Acarbose can potentiate the hypoglycemic effects of sulfonylureas or insulin.

Acarbose (BAY g 5421), antihyperglycemic agent, is an orally active alpha-glucosidase inhibitor (IC50=11 nM). Acarbose can potentiate the hypoglycemic effects of sulfonylureas or insulin.

Specifications

Chemical Name or Material Acarbose
Physical Form Powder
CAS 56180-94-0
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C25H43NO18
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym BAY g 5421
Solubility Information H2O : 125 mg/mL (193.62 mM; Need ultrasonic)
SMILES OC[C@H]([C@]1(O[C@@H]2[C@H](O)[C@@H](O)[C@@H]([C@H](O2)C)N[C@]3(C=C(CO)[C@H]([C@@H]([C@H]3O)O)O)[H])[H])O[C@@H]([C@@H]([C@H]1O)O)O[C@@]([H])([C@@H]([C@H](C=O)O)O)[C@@H](CO)O
Molecular Weight (g/mol) 645.6
Formula Weight 645.6
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.