missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acarbose, 95%

A reversible alpha-glucosidase inhibitor

£98.29 - £193.00

Chemical Identifiers

CAS 56180-94-0
Molecular Formula C25H43NO18
Molecular Weight (g/mol) 645.608
MDL Number MFCD00869592
InChI Key XUFXOAAUWZOOIT-AKWDJIIVSA-N
Synonym acarbose
PubChem CID 131674886
IUPAC Name (3S,5S)-5-[(2R,3S,5S)-5-[(2R,3S,5S)-3,4-dihydroxy-6-methyl-5-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino]oxan-2-yl]oxy-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,4-triol
SMILES CC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)NC4C=C(C(C(C4O)O)O)CO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15434279
SDS View Documents
Thermo Scientific™
J61737.03
1g
ONLINE EXCLUSIVE PRICE
£100.00
£98.29 / 1g
Save £1.71
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15444279
SDS View Documents
Thermo Scientific™
J61737.04
2g
ONLINE EXCLUSIVE PRICE
£153.00
£150.00 / 2g
Save £3.00
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15454279
SDS View Documents
Thermo Scientific™
J61737.06
5g
ONLINE EXCLUSIVE PRICE
£197.00
£193.00 / 5g
Save £4.00
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

56180-94-0
645.608
XUFXOAAUWZOOIT-AKWDJIIVSA-N
131674886
CC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)NC4C=C(C(C(C4O)O)O)CO
C25H43NO18
MFCD00869592
acarbose
(3S,5S)-5-[(2R,3S,5S)-5-[(2R,3S,5S)-3,4-dihydroxy-6-methyl-5-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino]oxan-2-yl]oxy-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,4-triol

Specifications

56180-94-0
MFCD00869592
acarbose
XUFXOAAUWZOOIT-AKWDJIIVSA-N
(3S,5S)-5-[(2R,3S,5S)-5-[(2R,3S,5S)-3,4-dihydroxy-6-methyl-5-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino]oxan-2-yl]oxy-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,4-triol
131674886
95%
C25H43NO18
14,18
Soluble in water.
CC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)NC4C=C(C(C(C4O)O)O)CO
645.608
645.61
Acarbose
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 260-030-7

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.