missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17570-1-AP-150UL
This item is not returnable.
View return policy
Description
ACAP3 is GTPase-activating protein for the ADP ribosylation factor family.Specifications
| ACAP3 | |
| Polyclonal | |
| Unconjugated | |
| ACAP3 | |
| ACAP3, Centaurin beta 5, CENTB5, Cnt b5, KIAA1716 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116983, 140500, 313772 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96P50 | |
| Acap3 | |
| ACAP3 Fusion Protein Ag11777 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction