missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAP1 Mouse anti-Human, Clone: 1B4C9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66596-1-IG-150UL
This item is not returnable.
View return policy
Description
ACAP1, also known as Centaurin-beta-1 (CENTB1), is a 740 amino acid containing transport carrier belonging to the Centaurin family with three ANK repeats, an Arf-GAP domain, a BAR domain and a PH domain that binds to phospholipids. ACAP1, a GTPase-activating protein (GAP) for ADP ribosylation factor 6 (ARF6), is required for clathrin-dependent export of proteins from recycling endosomes to trans-Golgi network and cell surface. It mediates CENTB1-binding to phosphatidylinositol 3,5-bisphosphate (PIP2) or phosphatidylinositol 3,4,5-trisphosphate (PIP3) containing membranes. Along with ACAP2, ACAP1 is recruited to platelet-derived growth factor (PDGF) induced dorsal membrane ruffles in NIH 3T3 mouse fibroblasts, while over expression inhibited ruffle formation. It is widely expressed in most tissues with high expression in lung and spleen and low expression in heart, kidney, liver and pancreas.Specifications
| ACAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15027 | |
| ACAP1 | |
| ACAP1 Fusion Protein Ag14254 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1B4C9 | |
| Unconjugated | |
| ACAP1 | |
| ACAP1, Centaurin beta 1, CENTB1, Cnt b1, KIAA0050 | |
| Mouse | |
| Protein G | |
| RUO | |
| 9744 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction