missing translation for 'onlineSavingsMsg'
Learn More

Acamprosate EP Impurity A, Mikromol™

Product Code. 30463066
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463066 4 x 25 mg -
30049393 25 mg 25mg
30051957 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30463066 Supplier Mikromol Supplier No. MM3407.0100254AMP
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Acamprosate EP Impurity A, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Aminopropane-1-sulfonic Acid (Homotaurine)
CAS 3687-18-1
Analyte or Component Names 3-Aminopropane-1-sulfonic Acid (Homotaurine)
Shipping Condition Room Temperature
Molecular Formula C3 H9 N O3 S
Certifications/Compliance ISO 17025
Synonym Acamprosate Calcium Imp. A (EP), Acamprosate Imp. A (EP), Acamprosate USP Related Compound A, Acamprosate USP RC A, 3-Aminopropane-1-sulfonic Acid, Homotaurine, Acamprosate Related Compound A, Acamprosate Calcium Impurity A, Acamprosate Impurity A
InChI Formula InChI=1S/C3H9NO3S/c4-2-1-3-8(5,6)7/h1-4H2,(H,5,6,7)
Solution Type Neat
SMILES NCCCS(=O)(=O)O
IUPAC Name 3-aminopropane-1-sulfonic acid
Molecular Weight (g/mol) 139.17
Quantity 4 x 25 mg
Formula Weight 139.0303 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.