missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acamprosate calcium, MedChemExpress
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Acamprosate calcium(Campral EC) is a GABA receptor agonist and modulator of glutamatergic systems
Specifications
Specifications
| Chemical Name or Material | Acamprosate calcium |
| CAS | 77337-73-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C5H10NO4S.1/2Ca |
| Quantity | 50 mg |
| Synonym | Calcium N-acetylhomotaurinate |
| Solubility Information | H2O : 100 mg/mL (499.40 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| SMILES | CC(NCCCS([O-])(=O)=O)=O.[0.5Ca2+] |
| Molecular Weight (g/mol) | 200.24 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction