missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACADS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100056
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ACADS.
Introduces a double bond at position 2 in saturated acyl-CoA's of short chain length, i. e. less than 6 carbon atoms.
Specifications
| ACADS | |
| Polyclonal | |
| Unconjugated | |
| ACADS | |
| ACAD3; Acads; acetyl-Coenzyme A dehydrogenase, short chain; acyl-CoA dehydrogenase short chain; acyl-CoA dehydrogenase, C-2 to C-3 short chain; acyl-Coenzyme A dehydrogenase, C-2 to C-3 short chain; acyl-Coenzyme A dehydrogenase, short chain; AI196007; Bcd1; Bcd-1; butyryl-CoA dehydrogenase; EGK_04272; Hdlq8; mitochondrial short-chain specific acyl-CoA dehydrogenase; SCAD; short chain acyl-coenzyme A dehydrogenase; short-chain acyl-CoA dehydrogenase; short-chain specific acyl-CoA dehydrogenase, mitochondrial; unsaturated acyl-CoA reductase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11409, 35, 64304 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P15651, P16219, Q07417 | |
| ACADS | |
| A synthesized peptide derived from human ACADS(Accession P16219), corresponding to amino acid residues I364-S412. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction