missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACADS Monoclonal Antibody (OTI1D2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526440
This item is not returnable.
View return policy
Description
ACADS Monoclonal Antibody for Western Blot, IHC (P)
Introduces a double bond at position 2 in saturated acyl-CoA's of short chain length, i. e. less than 6 carbon atoms.
Specifications
| ACADS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16219 | |
| ACADS | |
| Human recombinant protein fragment corresponding to amino acids 34-260 of ACADS produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D2 | |
| Unconjugated | |
| ACADS | |
| ACAD3; Acads; acetyl-Coenzyme A dehydrogenase, short chain; acyl-CoA dehydrogenase short chain; acyl-CoA dehydrogenase, C-2 to C-3 short chain; acyl-Coenzyme A dehydrogenase, C-2 to C-3 short chain; acyl-Coenzyme A dehydrogenase, short chain; AI196007; Bcd1; Bcd-1; butyryl-CoA dehydrogenase; EGK_04272; Hdlq8; mitochondrial short-chain specific acyl-CoA dehydrogenase; SCAD; short chain acyl-coenzyme A dehydrogenase; short-chain acyl-CoA dehydrogenase; short-chain specific acyl-CoA dehydrogenase, mitochondrial; unsaturated acyl-CoA reductase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 35 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction