missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACADM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55210-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to ACADM.
This gene encodes the medium-chain specific (C4 to C12 straight chain) acyl-Coenzyme A dehydrogenase. The homotetramer enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Defects in this gene cause medium-chain acyl-CoA dehydrogenase deficiency, a disease characterized by hepatic dysfunction, fasting hypoglycemia, and encephalopathy, which can result in infantile death. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| ACADM | |
| Polyclonal | |
| Unconjugated | |
| ACADM | |
| ACAD1, ACADM, MCAD, MCADH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11364, 24158, 34 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08503, P11310, P45952 | |
| ACADM | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction