missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACADM Polyclonal Antibody, Biotin

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142019
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognise both reported isoforms (NP_000007.1; NP_001120800.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
This gene encodes the medium-chain specific (C4 to C12 straight chain) acyl-Coenzyme A dehydrogenase. The homotetramer enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Defects in this gene cause medium-chain acyl-CoA dehydrogenase deficiency, a disease characterized by hepatic dysfunction, fasting hypoglycemia, and encephalopathy, which can result in infantile death. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| ACADM | |
| Polyclonal | |
| Biotin | |
| ACADM | |
| ACAD1; Acadm; acyl-CoA dehydrogenase C-4 to C-12 straight chain; acyl-CoA dehydrogenase, C-4 to C-12 straight chain; Acyl-Coenzyme A dehydrogenase C-4 to C-12 straight-chain; acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight chain; Acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight-chain; acyl-Coenzyme A dehydrogenase, medium chain; AU018656; FLJ18227; FLJ93013; FLJ99884; MCAD; MCADH; medium-chain acyl-CoA dehydrogenase; medium-chain specific acyl-CoA dehydrogenase, mitochondrial; RP4-682C21.1; testicular tissue protein Li 7 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 34 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P11310 | |
| ACADM | |
| Peptide with sequence C-RLIVAREHIDKYKN (aa 408-421). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction