missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACADM Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67742-1-IG-20UL
This item is not returnable.
View return policy
Description
The ACADM antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ACADM. This antibody recognizes human, rat, pig antigen. The ACADM antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.Specifications
| ACADM | |
| Monoclonal | |
| 1000 μg/mL | |
| ACAD1; FLJ18227; FLJ93013; FLJ99884; MCAD; MCADH | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Rat, Pig | |
| 42 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1A11G6 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 34 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur