missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACADL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540791
This item is not returnable.
View return policy
Description
Peptide sequence: MAARLLRGSL RVLGGHRAPR QLPAARCSHS GGEERLETPS AKKLTDIGIR Sequence homology: Cow: 93%; Dog: 86%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 93%; Rabbit: 85%; Rat: 92%.
The protein encoded by this gene belongs to the acyl-CoA dehydrogenase family, which is a family of mitochondrial flavoenzymes involved in fatty acid and branched chain amino-acid metabolism. This protein is one of the four enzymes that catalyze the initial step of mitochondrial beta-oxidation of straight-chain fatty acid. Defects in this gene are the cause of long-chain acyl-CoA dehydrogenase (LCAD) deficiency, leading to nonketotic hypoglycemia.
Specifications
| ACADL | |
| Polyclonal | |
| Unconjugated | |
| ACADL | |
| AA960361; ACAD4; ACADL; acetyl-Coenzyme A dehydrogenase, long-chain; ACOADA; Acyl Coenzyme A dehydrogenase, long chain; acyl-CoA dehydrogenase long chain; acyl-CoA dehydrogenase, long chain; acyl-Coenzyme A dehydrogenase, long chain; acyl-Coenzyme A dehydrogenase, long-chain; AU018452; C79855; LCAD; LCAD long chain acyl-CoA dehydrogenase; LOC100052267; long-chain acyl-CoA dehydrogenase; long-chain specific acyl-CoA dehydrogenase, mitochondrial; uncharacterized protein LOC100052267; zgc:55656 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 33 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P28330 | |
| ACADL | |
| Synthetic peptide directed towards the N-terminal of human ACADL (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction