missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAD9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £368.00
Specifications
| Antigen | ACAD9 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600142
|
Novus Biologicals
NBP2-92735-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625922
|
Novus Biologicals
NBP2-92735-0.1ml |
0.1 mL |
£368.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACAD9 Polyclonal antibody specifically detects ACAD9 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ACAD9 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 28976 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ACAD-9, acyl-CoA dehydrogenase family member 9, mitochondrial, acyl-CoA dehydrogenase family, member 9, acyl-Coenzyme A dehydrogenase family, member 9, EC 1.3.99, EC 1.3.99.-, EC 1.3.99.10, FLJ23533, MGC14452, NPD002 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human ACAD9 (NP_054768.2). MSGCGLFLRTTAAARACRGLVVSTANRRLLRTSPPVRAFAKELFLGKIKKKEVFPFPEVSQDELNEINQFLGPVEKFFTEEVDSRKIDQEGKIPDETLEKLKSLGLFGLQVPEEYGGLGFSNTMYSRLGEIISMDGSITVTLAAHQAIGLKGIILAGTEEQKAKYLPKLASGEHIAAFCLTEPASGSDAASIRSRATLSEDKKHYILNGSKVWITNGGLANIFTVFAKTEVVDSDGSVKDKITAFIVERDFGGVTNGKPEDKLGIRGSNT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title