missing translation for 'onlineSavingsMsg'
Learn More

ACAD11 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18318872 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18318872 25 μg 25µg
18385783 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18318872 Supplier Novus Biologicals Supplier No. NBP31191725UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ACAD11 Polyclonal antibody specifically detects ACAD11 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAD11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS
Gene Alias ACAD-11, acyl-CoA dehydrogenase family member 11, acyl-CoA dehydrogenase family, member 11, acyl-Coenzyme A dehydrogenase family, member 11, EC 1.3.99.-, FLJ12592, MGC150619
Host Species Rabbit
Immunogen Synthetic peptide derived from the human ACAD11 protein (Uniprot: Q709F0)
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84129
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.