missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAD11 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £359.00
Specifications
| Antigen | ACAD11 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637120
|
Novus Biologicals
NBP2-92304-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658181
|
Novus Biologicals
NBP2-92304-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACAD11 Polyclonal antibody specifically detects ACAD11 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ACAD11 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 84129 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ACAD-11, acyl-CoA dehydrogenase family member 11, acyl-CoA dehydrogenase family, member 11, acyl-Coenzyme A dehydrogenase family, member 11, EC 1.3.99.-, FLJ12592, MGC150619 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human ACAD11 (NP_115545.3). MKPGATGESDLAEVLPQHKFDSKSLEAYLNQHLSGFGAEREATLTIAQYRAGKSNPTFYLQKGFQTYVLRKKPPGSLLPKAHQIDREFKVQKALFSIGFPVPKPILYCSDTSVIGTEFYVMEHVQGRIFRDLTIPGLSPAERSAIYVATVETLAQLHSLNIQSLQLEGYGIGAGYCKRQVSTWTKQYQAAAHQDIPAMQQLSEWLMKNLPDNDNEENLIHGDFRLDNIVF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title