missing translation for 'onlineSavingsMsg'
Learn More

Acacetin, MedChemExpress

Product Code. 16483866
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483866 25 mg 25mg
16468988 10 mg 10mg
16401418 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16483866 Supplier Medchem Express Supplier No. HYN0451
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Acacetin (5,7-Dihydroxy-4'-methoxyflavone) is an orally active flavonoid derived from Tephroseris kirilowii (Turcz.) Holub. Acacetin docks in the ATP binding pocket of PI3Kγ. Acacetin causes cell cycle arrest and induces apoptosis and autophagy in cancer cells. Acacetin has potent anti-cancer and anti-inflammatory activity and has the potential for pain-related diseases research.

Acacetin (5,7-Dihydroxy-4'-methoxyflavone) is an orally active flavonoid derived from Tephroseris kirilowii (Turcz.) Holub. Acacetin docks in the ATP binding pocket of PI3Kγ. Acacetin causes cell cycle arrest and induces apoptosis and autophagy in cancer cells. Acacetin has potent anti-cancer and anti-inflammatory activity and has the potential for pain-related diseases research.

Specifications

Chemical Name or Material Acacetin
CAS 480-44-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H12O5
Quantity 25 mg
Synonym 5,7-Dihydroxy-4'-methoxyflavone
Solubility Information DMSO : ≥ 37 mg/mL (130.16 mM)
SMILES O=C1C=C(C2=CC=C(OC)C=C2)OC3=CC(O)=CC(O)=C13
Molecular Weight (g/mol) 284.26
Formula Weight 284.26
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.