missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACACB Recombinant Rabbit Monoclonal Antibody (K01_1M31)
Description
ACACB Recombinant Monoclonal Antibody for Western Blot
Catalyzes the ATP-dependent carboxylation of acetyl-CoA to malonyl-CoA. Carries out three functions: biotin carboxyl carrier protein, biotin carboxylase and carboxyltransferase. Involved in inhibition of fatty acid and glucose oxidation and enhancement of fat storage. May play a role in regulation of mitochondrial fatty acid oxidation through malonyl-CoA-dependent inhibition of carnitine palmitoyltransferase 1.
Specifications
Specifications
| Antigen | ACACB |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1M31 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | ACACB |
| Gene Accession No. | O00763 |
| Gene Alias | Acacb; Acc2; ACCB; ACC-beta; acetyl-CoA carboxylase 2; acetyl-CoA carboxylase beta; acetyl-Coenzyme A carboxylase 2; acetyl-Coenzyme A carboxylase beta; AI597064; AW743042; Biotin carboxylase; HACC275 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?