missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACACB Recombinant Rabbit Monoclonal Antibody (24GB3230)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556949
This item is not returnable.
View return policy
Description
ACACB Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Catalyzes the ATP-dependent carboxylation of acetyl-CoA to malonyl-CoA. Carries out three functions: biotin carboxyl carrier protein, biotin carboxylase and carboxyltransferase. Involved in inhibition of fatty acid and glucose oxidation and enhancement of fat storage. May play a role in regulation of mitochondrial fatty acid oxidation through malonyl-CoA-dependent inhibition of carnitine palmitoyltransferase 1.
Specifications
| ACACB | |
| Recombinant Monoclonal | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| E9Q4Z2, O00763 | |
| ACACB | |
| A synthesized peptide derived from human Acetyl-CoA Carboxylase (aa 350-450). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3230 | |
| Unconjugated | |
| ACACB | |
| Acacb; Acc2; ACCB; ACC-beta; acetyl-CoA carboxylase 2; acetyl-CoA carboxylase beta; acetyl-Coenzyme A carboxylase 2; acetyl-Coenzyme A carboxylase beta; AI597064; AW743042; Biotin carboxylase; HACC275 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100705, 116719, 32 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction