missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAA1 Antibody (CL2650) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42380JF669
This item is not returnable.
View return policy
Description
ACAA1 Monoclonal antibody specifically detects ACAA1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| ACAA1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Acetyl-CoA acyltransferase, acetyl-CoA acyltransferase 1, EC 2.3.1, peroxisomal | |
| This antibody was developed using a recombinant protein derived from P09110, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Proteases & Other Enzymes, Signal Transduction | |
| 30 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL2650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction