missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ac4ManNAz
GREENER_CHOICE

Product Code. 18749016
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18749016

Brand: Tocris Bioscience™ 7479/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Azide-containing monosaccharide building block; used as glycoprotein, glycolipid and glycoRNA probe

Ac4ManNAz is a cell-permeable, unnatural azide-containing monosaccharide building block. Ac4ManNAz is metabolized and incorporated as azidosialic acid into cell surface and secreted sialoglycoproteins to allow Cu(I)-free click chemistry for further conjugation or detection. Ac4ManNAz can be used for bioorthogonal labeling of human prostate tissue slice cultures; the target sialoglycoproteins can be biotinylated for proteomic analysis or reacted with a cyclooctyne-functionalized optical probe for imaging. Ac4ManNAz is also used to label glycoRNAs on the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(2-Azidoacetyl)amino]-2-deoxy-D-mannopyranose-1,3,4,6-tetraacetate
CAS 361154-30-5
Quantity 10 mg
Target Photoactivatable Cross-linking Probes
Molecular Formula C16H22N4O10
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.