missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ac4GalN6yne
GREENER_CHOICE

Product Code. 30000444
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30000444

Brand: Tocris Bioscience™ 7932/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alkyne containing monosaccharide building block; used in BOCTAG

Ac4GalN6yne is a cell-permeable alkyne-containing monosaccharide building block. It can be used in Bio-Orthogonal Cell-specific Tagging of Glycoproteins (BOCTAG) technique. In this strategy, cells carrying an artificial biosynthetic pathway (B. longum NahK and mut-AGX1) can generate alkyne-tagged UDP-GalN6yne, that are then used by glycosyltransferases to chemically tag the glycoproteome. Ac4GalN6yne can be used for bioorthogonal labelling in vitro and mouse models.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Deoxy-2-[(1-oxo-5-hexyn-1-yl)amino]-D-galactopyranose-1,3,4,6-tetraacetate
Quantity 10 mg
Target Glycobiology Probes
Molecular Formula C20H27NO10
Purity 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.