missing translation for 'onlineSavingsMsg'
Learn More

AC1 Antibody (3F12), Novus Biologicals™

Product Code. 18420140 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420140 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420140 Supplier Novus Biologicals Supplier No. H00010141M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AC1 Monoclonal antibody specifically detects AC1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen AC1
Applications ELISA
Classification Monoclonal
Clone 3F12
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005741
Gene Alias aC1, chromosome 4 open reading frame 6, expressed in neuroblastoma, Protein AC1
Host Species Mouse
Immunogen C4orf6 (NP_005741, 1 a.a. ~ 93 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDTQKQIHKTHNSKNQFFTIFFFLSVEFGKEGTRKNFYLLLSIGHYGRKSRRADLGTADTADKTEPECFAASWTFDPNPSVTVSGAHSTAVHQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10141
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.