missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure™ Ac-rC Phosphoramidite, 28-400 finish bottle

Product Code. 17947182 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17947182 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17947182 Supplier Thermo Scientific™ Supplier No. 27180513
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ac-rC CEP is a phosphoramidite containing 5' DMT and the nucleoside cytidine (C) with Acetyl (Ac) base protection, for use in oligonucleotide synthesis. RNA phosphoramidites contain a 2'-OH protecting group, tBDMS (tert-butyl dimethylsilyl).

Ac-rC CEP is a phosphoramidite containing 5' DMT and the nucleoside cytidine (C) with Acetyl (Ac) base protection, for use in oligonucleotide synthesis. RNA phosphoramidites contain a 2'-OH protecting group, tBDMS (tert-butyl dimethylsilyl). Each base has unique protecting groups designed specifically to protect the RNA during the chemical synthesis process, and produce the highest amount of full length product. Alternate base protection options are also available to allow for faster deprotection and longer oligonucleotide synthesis. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 30 mL Amber bottle with a 28-400 Finish neck style, which is compatible with most MerMade Systems of oligonucleotide synthesizers.

Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N4-acetyl-2'-O-[(tert-butyl)dimethylsilyl]-cytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C47H64N5O9PSi
• Base: rC
• Base Protection: Ac
• Formula weight: 902.1 g/mol
• CAS number: 121058-88-6
• HPLC Purity: ≥99%; M-11 Impurity: ≤0.01%
• 31P NMR Purity: ≥99%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%; No single PIII impurity >0.3%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) MerMade Synthesizers
Product Type Ac-rC CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Cytidine (C)
Reagent Features Fast deprotection
Shipping Condition Room Temperature
Purity 99%
Quantity 2 g
Reagent Type Phosphoramidite
Grade TheraPure™
Nucleic Acid Type RNA
Product Line TheraPure
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.