Learn More
Invitrogen™ ac Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569771
Description
This target displays homology in the following species: Human: 79%; Rat: 85% Peptide sequence: FNGPSVIRRNARERNRVKQVNNGFSQLRQHIPAAVIADLSNGRRGIGPGA For short term use, store at 2-8C up to 1 week. For long term storage, store at -20C in small aliquots to prevent freeze-thaw cycles. Concentration is batch dependent: 0.5-1 mg/mL.
AS-C proteins are involved in the determination of the neuronal precursors in the peripheral nervous system and the central nervous system.
Specifications
| ac | |
| Polyclonal | |
| Unconjugated | |
| ac | |
| 990 E5 F1; Ac; achaete; Achaete/Scute; achaete-scute; Achaete-scute complex protein T5; acheate; achete; ac-PA; ASC; AS-C; AS-C T5; AS-C T5ac; ascT5; CG3796; CG3796-PA; Dmel\CG3796; Dmel_CG3796; EG:125H10.3; hairy wing; Hairy-wing; Hw; IP01413p; Protein achaete; sc/T5; T5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 30981 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P10083 | |
| ac | |
| synthetic peptide corresponding to a region of fruit fly ac (aa 21-70). | |
| 100 μL | |
| Primary | |
| Drosophila | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.