missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Ac-Leu-Arg-AMC

Product Code. 16414056
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mg
missing translation for 'unitSize'
25mg
This item is not returnable. View return policy

Product Code. 16414056

missing translation for 'mfr': MedChemExpress HYP1448

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ac-Leu-Arg-AMC is a fluorogenic peptide substrate.

Specifications

CAS 929621-79-4
Solubility DMSO : 300 mg/mL (616.57 mM∣Need ultrasonic and warming)
Content And Storage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Neuroscience-Neuromodulation
Quantity 25 mg
Molecular Weight (g/mol) 486.56 g/mol
Product Type Dye Reagents
Purity or Quality Grade 99.28%
Shipping Condition Room temperature
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.