missing translation for 'onlineSavingsMsg'
Learn More

Ac-CoA Synthase Inhibitor1, MedChemExpress

Product Code. 16472916
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16472916

Brand: Medchem Express HY104032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ac-CoA Synthase Inhibitor1 is a potent, reversible acetate-dependent acetyl-CoA synthetase 2 (ACSS2) inhibitor with an IC50 of 0.6 μM. Ac-CoA Synthase Inhibitor1 inhibits the respiratory syncytial virus (RSV).

Ac-CoA Synthase Inhibitor1 is a potent, reversible acetate-dependent acetyl-CoA synthetase 2 (ACSS2) inhibitor with an IC50 of 0.6 μM. Ac-CoA Synthase Inhibitor1 inhibits the respiratory syncytial virus (RSV).

Specifications

Chemical Name or Material Ac-CoA Synthase Inhibitor1
Physical Form Powder
CAS 508186-14-9
Color Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C20H18N4O2S2
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 31.25 mg/mL (76.12 mM; Need ultrasonic)
SMILES O=C(NCCOC)NC1=CC=C2N=C(C3=CC=CS3)C(C4=CC=CS4)=NC2=C1
Molecular Weight (g/mol) 410.51
Formula Weight 410.51
Percent Purity 98.56%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.