missing translation for 'onlineSavingsMsg'
Learn More

Brady™ AC Adapter

Product Code. 11776365
Click to view available options
Color:
Black
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11776365

Brand: Brady™ undefined

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

AC Adapter for the BMP™51/53 Label Makers.

  • Accessories for the BMP™51/53 Label Makers.

Specifications

Product Type AC Adapter
Color Black
For Use With (Equipment) BMP41 Label Printer, BMP51 Label Printer, BMP53 Label Printer, BMP71 Label Printer, BradyPrinter M611
Description AC Adaptor, USA 120 V
Format AC Adapter
Quantity 1 Ea.
Voltage 120 V
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.