missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Universal AC Adapter

Product Code. 16393677 Shop All Brand Products
Click to view available options
:
Universal AC Adapter
This item is not returnable. View return policy

Product Code. 16393677

Brand: BRANDTECH™ BRAND™ 705250

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with HandyStep™ touch and HandyStep™ touch S.

  • Includes USB cable and 5 country adapters
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, C/US
Autoclavable Not Autoclavable
Color Black
For Use With BRAND HandyStep Touch/Touch S
Material Plastic
Type AC Adapter/Power Supply
For Use With (Equipment) HandyStep Touch / Touch S
Plug Type US, UK, CEE, CN
Quantity 1
Electrical Requirements 100 to 240 V, 50/60 Hz
Amperage 0.5 A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.