missing translation for 'onlineSavingsMsg'
Learn More

AC-55649, MedChemExpress

Product Code. 16434208
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434208 10 mg 10mg
16412996 25 mg 25mg
16406927 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16434208 Supplier Medchem Express Supplier No. HY108526

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AC-55649 is a potent, highly isoform-selective agonist of human RARβ2 receptor, with a pEC50 of 6.9.

AC-55649 is a potent, highly isoform-selective agonist of human RARβ2 receptor, with a pEC50 of 6.9.

Specifications

Chemical Name or Material AC-55649
Physical Form Solid
CAS 59662-49-6
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H26O2
Solubility Information DMSO : 100 mg/mL (322.13 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C2=CC=C(CCCCCCCC)C=C2)C=C1)O
Molecular Weight (g/mol) 310.43
Formula Weight 310.43
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.