missing translation for 'onlineSavingsMsg'
Learn More

Ravoxertinib hydrochloride, MedChemExpress

Product Code. 16406928
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16406928

Brand: Medchem Express HY15947A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ravoxertinib hydrochloride (GDC-0994 hydrochloride) is an orally bioavailable inhibitor selective for ERK kinase activity with IC50 of 6.1 nM and 3.1 nM for ERK1 and ERK2, respectively.

Ravoxertinib hydrochloride (GDC-0994 hydrochloride) is an orally bioavailable inhibitor selective for ERK kinase activity with IC50 of 6.1 nM and 3.1 nM for ERK1 and ERK2, respectively.

Specifications

Chemical Name or Material Ravoxertinib hydrochloride
CAS 2070009-58-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H19Cl2FN6O2
Quantity 10 mg
Synonym GDC-0994 hydrochloride
Solubility Information DMSO : 100 mg/mL (209.50 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 80°C) (insoluble)
SMILES O=C1C=C(C2=NC(NC3=CC=NN3C)=NC=C2)C=CN1[C@@H](C4=CC=C(Cl)C(F)=C4)CO.Cl[H]
Molecular Weight (g/mol) 477.32
Formula Weight 477.32
Percent Purity 98.99%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.