missing translation for 'onlineSavingsMsg'
Learn More

AC-55541, MedChemExpress

Product Code. 16458647
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16458647

Brand: Medchem Express HY143505mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AC-55541 is a highly selective protease-activated receptor 2 (PAR2) agonist (pEC50=6.7), displays no activity at other PAR subtypes or at over 30 other receptors involved in nociception and inflammation. AC-55541 has pEC50 values of 5.9 and 6.6 in PI hydrolysis assays and Ca2+ mobilization assays and exhibits pronociceptive activity in vivo.

AC-55541 is a highly selective protease-activated receptor 2 (PAR2) agonist (pEC50=6.7), displays no activity at other PAR subtypes or at over 30 other receptors involved in nociception and inflammation. AC-55541 has pEC50 values of 5.9 and 6.6 in PI hydrolysis assays and Ca2+ mobilization assays and exhibits pronociceptive activity in vivo.

Specifications

Chemical Name or Material AC-55541
CAS 916170-19-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H20BrN5O3
Quantity 5 mg
Solubility Information DMSO : ≥ 51 mg/mL (98.39 mM)
SMILES O=C(NC(C(C1=CC=CC=C12)=NNC2=O)C(N/N=C(C)/C3=CC=CC(Br)=C3)=O)C4=CC=CC=C4
Molecular Weight (g/mol) 518.36
Formula Weight 518.36
Percent Purity 99.19%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.